Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnj11
Entrez Gene ID
16514
Gene Name
potassium inwardly rectifying channel, subfamily J, member 11
Gene Aliases
Kir6.2, mBIR
Chromosome Location
Chr.7: 46097123 - 46100872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010602.3 NP_034732.1 1 - 1 447 66
GenBank mRNA BC052731.1 - 1 - 1 447 66
BC057006.1 - 1 - 1 447 66
D50581.1 - 1 - 1 238 66
U73626.1 - 1 - 1 361 66

View Full Product Details