Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk14
Entrez Gene ID
317653
Gene Name
kallikrein related-peptidase 14
Gene Aliases
GK14
Chromosome Location
Chr.7: 43680879 - 43695535 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174866.2 NP_777355.1 1 - 2 45 97
GenBank mRNA BC128019.1 - 1 - 2 48 97

View Full Product Details