The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204875.1 | NP_001191804.1 | 8 - 8 | 2289 | 113 |
NM_023871.4 | NP_076360.1 | 8 - 8 | 2154 | 113 | |
GenBank mRNA | AK019960.1 | - | 8 - 8 | 2040 | 113 |
AK150304.1 | - | 1 - 1 | 947 | 113 | |
AK172327.1 | - | 8 - 8 | 2297 | 113 | |
BC012508.1 | - | 2 - 2 | 1229 | 113 |