The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_015385.2 | 2 - 3 | 582 | 102 | |
GenBank mRNA | AY590889.1 | - | 2 - 3 | 410 | 102 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | calmodulin-lysine N-methyltransferase | Camkmt | 1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT | -150.5kb | Anti-sense | Chr. - 85090592 - 85458589 on Build GRCm38 |
Downstream | sine oculis-related homeobox 3 | Six3 | E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta | 5.0kb | Anti-sense | Chr. - 85620834 - 85626191 on Build GRCm38 |