Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- B130034C11Rik
- Entrez Gene ID
-
414095
- Gene Name
- RIKEN cDNA B130034C11 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 87496073 - 87504038 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040375.1 |
|
2 - 3 |
576 |
104 |
GenBank mRNA |
AK045112.1 |
- |
2 - 3 |
577 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chaperonin containing Tcp1, subunit 8 (theta) |
Cct8 |
AI132397, Cctq, Tcpq |
-6.0kb
|
Anti-sense |
Chr. - 87483325 - 87495869 on Build GRCm38 |
Downstream |
Map3k7 C-terminal like |
Map3k7cl |
C21orf7, ORF63, Tak1l |
49.3kb
|
Sense |
Chr. - 87553330 - 87595336 on Build GRCm38 |
View Full Product Details