Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mospd4
- Entrez Gene ID
-
72076
- Gene Name
- motile sperm domain containing 4
- Gene Aliases
-
2010013H21Rik
- Chromosome Location
-
Chr.18: 46464934 - 46465816 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045438.1 |
|
1 - 1 |
825 |
86 |
GenBank mRNA |
AK076850.1 |
- |
1 - 1 |
767 |
86 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV207619.1 |
- |
1 |
177 |
|
Chr. 18 - 46464934 - 46465169 [-] on Build GRCm38 |
86 |
GenBankmRNA |
AV208876.1 |
- |
1 |
240 |
|
Chr. 18 - 46464934 - 46465229 [-] on Build GRCm38 |
86 |
GenBankmRNA |
AV265561.2 |
- |
1 |
573 |
|
Chr. 18 - 46464934 - 46465563 [-] on Build GRCm38 |
86 |
GenBankmRNA |
BX632683.1 |
- |
1 |
|
|
Chr. 18 - 46464925 - 46465153 [+] on Build GRCm38 |
86 |
GenBankmRNA |
AV209040.1 |
- |
1 |
236 |
|
Chr. 18 - 46464934 - 46465207 [-] on Build GRCm38 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene 4107 |
Gm4107 |
|
-37.3kb
|
Sense |
Chr. - 46357872 - 46427619 on Build GRCm38 |
Downstream |
fem-1 homolog c (C.elegans) |
Fem1c |
2610312A07Rik, 3632443A22Rik, AI646567 |
38.8kb
|
Sense |
Chr. - 46504606 - 46525971 on Build GRCm38 |
View Full Product Details