Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etohi1
Entrez Gene ID
626848
Gene Name
ethanol induced 1
Gene Aliases
-
Chromosome Location
Chr.2: 178023284 - 178035859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177399.1 NP_001170870.1 4 - 4 2219 125

View Full Product Details