Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cwc22
Entrez Gene ID
80744
Gene Name
CWC22 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
AA684037, AI173004, AL022752, B230213M24, mKIAA1604
Chromosome Location
Chr.2: 77895653 - 77946356 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290740.1 NP_001277669.1 5 - 6 757 65
NM_030560.5 NP_085037.2 6 - 7 815 65
NM_172667.2 NP_766255.1 5 - 6 754 65
GenBank mRNA AK045589.1 - 5 - 6 586 65
AK051237.1 - 5 - 6 582 65
AK077961.1 - 6 - 7 643 65
AK089988.1 - - 593 65
AK129404.1 - - 571 65
AK132001.1 - 5 - 6 698 65
AK143352.1 - 5 - 6 587 65
AK149528.1 - 5 - 6 581 65
AK166487.1 - 5 - 6 593 65
BC003993.1 - 5 - 6 567 65

View Full Product Details