Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Alg9
- Entrez Gene ID
-
102580
- Gene Name
- asparagine-linked glycosylation 9 (alpha 1,2 mannosyltransferase)
- Gene Aliases
-
8230402H15Rik, AI747665, B430313H07Rik, Dibd1
- Chromosome Location
-
Chr.9: 50774881 - 50843639 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045601.1 |
|
3 - 4 |
565 |
76 |
GenBank mRNA |
AK149592.1 |
- |
3 - 4 |
487 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CF723659.1 |
- |
2 - 3 |
236 |
|
Chr. 9 - 50776581 - 50800222 [+] on Build GRCm38 |
76 |
GenBankmRNA |
BQ572382.1 |
- |
1 - 2 |
101 |
|
Chr. 9 - 50779014 - 50802015 [+] on Build GRCm38 |
76 |
GenBankmRNA |
CJ134066.1 |
- |
3 - 4 |
487 |
|
Chr. 9 - 50775305 - 50789579 [+] on Build GRCm38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ferredoxin-fold anticodon binding domain containing 1 |
Fdxacb1 |
D630004A14Rik |
-6.3kb
|
Sense |
Chr. - 50768238 - 50772670 on Build GRCm38 |
Downstream |
protein phosphatase 2, regulatory subunit A, beta |
Ppp2r1b |
2410091N08Rik, AI790395 |
68.4kb
|
Sense |
Chr. - 50856615 - 50899313 on Build GRCm38 |
View Full Product Details