Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gm10640
Entrez Gene ID
100038516
Gene Name
predicted gene 10640
Gene Aliases
EG668322, ENSMUSG00000074193
Chromosome Location
Chr.7: 31162358 - 31170641 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_046062.1 2 - 3 540 76
GenBank mRNA AK133438.1 - 2 - 3 540 76


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ047870.1 - 2 - 3 540 Chr. 7 - 31162358 - 31163934 [+] on Build GRCm38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GRAM domain containing 1A Gramd1a 1300003M23Rik, AI415002, AW547240, D7Bwg0611e -7.5kb Anti-sense Chr. - 31130127 - 31155935 on Build GRCm38
Downstream secretoglobin, family 1B, member 2 Scgb1b2 Abpa2, Abph, Apbh, LGP, Mja1l, Scgb1b1 126.6kb Anti-sense Chr. - 31290519 - 31291816 on Build GRCm38

View Full Product Details