Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pglyrp2
- Entrez Gene ID
-
57757
- Gene Name
- peptidoglycan recognition protein 2
- Gene Aliases
-
C730002N09Rik, PGRP-L, Pglyrpl, tagL
- Chromosome Location
-
Chr.17: 32412459 - 32424183 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073184.1 |
|
2 - 3 |
1057 |
115 |
GenBank mRNA |
AF315584.1 |
- |
2 - 3 |
1057 |
115 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA A530088E08 gene |
A530088E08Rik |
|
-12.1kb
|
Anti-sense |
Chr. - 32403014 - 32404808 on Build GRCm38 |
Downstream |
cytochrome P450, family 4, subfamily f, polypeptide 39 |
Cyp4f39 |
4732474A20Rik |
33.7kb
|
Anti-sense |
Chr. - 32452695 - 32493320 on Build GRCm38 |
View Full Product Details