Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npm1
Entrez Gene ID
18148
Gene Name
nucleophosmin 1
Gene Aliases
B23, NO38, Npm
Chromosome Location
Chr.11: 33152498 - 33163206 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252260.1 NP_001239189.1 2 - 3 321 88
GenBank mRNA BU513690.1 - 2 - 3 177 88

View Full Product Details