Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bin2
Entrez Gene ID
668218
Gene Name
bridging integrator 2
Gene Aliases
-
Chromosome Location
Chr.15: 100641082 - 100669576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270537.1 NP_001257466.1 9 - 10 773 109

View Full Product Details