Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd12b
Entrez Gene ID
100504285
Gene Name
abhydrolase domain containing 12B
Gene Aliases
Gm804
Chromosome Location
Chr.12: 70154171 - 70183206 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195033.1 NP_001181962.1 11 - 12 935 80

View Full Product Details