Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd12b
Entrez Gene ID
100504285
Gene Name
abhydrolase domain containing 12B
Gene Aliases
Gm804
Chromosome Location
Chr.12: 70154171 - 70183206 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195033.1 NP_001181962.1 5 - 6 477 94

View Full Product Details