Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl3
Entrez Gene ID
100503085
Gene Name
kelch-like 3
Gene Aliases
7530408C15Rik, AI430941, EG627648
Chromosome Location
Chr.13: 58004439 - 58123481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195075.1 NP_001182004.1 14 - 15 1610 105

View Full Product Details