Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lce3d
Entrez Gene ID
630994
Gene Name
late cornified envelope 3D
Gene Aliases
ENSMUSG00000043472, Gm9782
Chromosome Location
Chr.3: 92957390 - 92958574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270426.1 NP_001257355.1 2 - 2 523 77

View Full Product Details