Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Klrb1-ps1
- Entrez Gene ID
-
724020
- Gene Name
- killer cell lectin-like receptor subfamily B member 1, pseudogene 1
- Gene Aliases
-
Nkrp1e
- Chromosome Location
-
Chr.6: 129116518 - 129129446 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073569.1 |
|
3 - 4 |
254 |
134 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
C-type lectin domain family 2, member e |
Clec2e |
Clra, clr-a |
-20.1kb
|
Anti-sense |
Chr. - 129091816 - 129100903 on Build GRCm38 |
Downstream |
C-type lectin domain family 2, member d |
Clec2d |
Clr-b, Clrb, Ocil |
57.5kb
|
Sense |
Chr. - 129180615 - 129186535 on Build GRCm38 |
View Full Product Details