Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd10
Entrez Gene ID
213012
Gene Name
abhydrolase domain containing 10
Gene Aliases
-
Chromosome Location
Chr.16: 45729720 - 45743278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073572.1 4 - 5 470 76
GenBank mRNA CX237340.1 - 4 - 5 417 76


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CX203392.1 - 4 - 5 404 Chr. 16 - 45731699 - 45742889 [-] on Build GRCm38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transgelin 3 Tagln3 2700038H05Rik, 2900005O10Rik, AI426007, Np25 -5.2kb Sense Chr. - 45711230 - 45724531 on Build GRCm38
Downstream pleckstrin homology-like domain, family B, member 2 Phldb2 AV253284, C820004H04Rik, LL5b, LL5beta 13.5kb Sense Chr. - 45746230 - 45953701 on Build GRCm38

View Full Product Details