The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_075094.1 | 2 - 3 | 1672 | 86 | ||
GenBank mRNA | AK044313.1 | - | LIT3405 Vax2OS |
2 - 3 | 1672 | 86 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | CD207 antigen | Cd207 | -24.3kb | Sense | Chr. - 83671207 - 83677857 on Build GRCm38 | |
Downstream | ventral anterior homeobox 2 | Vax2 | Dres93 | 7.2kb | Anti-sense | Chr. - 83711264 - 83739229 on Build GRCm38 |