Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rnf41
- Entrez Gene ID
-
67588
- Gene Name
- ring finger protein 41
- Gene Aliases
-
2210404G21Rik, 4930511A05Rik, 4933415P08Rik, D10Ertd722e, FLRF, Nrdp1
- Chromosome Location
-
Chr.10: 128411616 - 128441441 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104290.1 |
|
3 - 4 |
507 |
95 |
GenBank mRNA |
AK016825.1 |
- |
2 - 3 |
238 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY716499.1 |
- |
2 - 3 |
238 |
|
Chr. 10 - 128415178 - 128437418 [+] on Build GRCm38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
nucleic acid binding protein 2 |
Nabp2 |
2610036N15Rik, Obfc2b |
-17.1kb
|
Anti-sense |
Chr. - 128401394 - 128411487 on Build GRCm38 |
Downstream |
microRNA 8105 |
Mir8105 |
Gm27346, mmu-mir-8105 |
26.4kb
|
Sense |
Chr. - 128458680 - 128458768 on Build GRCm38 |
View Full Product Details