Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chchd7
Entrez Gene ID
66433
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 7
Gene Aliases
1810049H20Rik
Chromosome Location
Chr.4: 3938860 - 3951382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104355.1 6 - 7 995 87
GenBank mRNA AK145105.1 - 6 - 7 970 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream pleiomorphic adenoma gene 1 Plag1 -7.2kb Anti-sense Chr. - 3898769 - 3938434 on Build GRCm38
Downstream short chain dehydrogenase/reductase family 16C, member 5 Sdr16c5 Rdhe2, Scdr9 48.8kb Anti-sense Chr. - 3995942 - 4019663 on Build GRCm38

View Full Product Details