Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo44
Entrez Gene ID
230903
Gene Name
F-box protein 44
Gene Aliases
5730411K09, AV001623, FBG3, FBX30, Fbx6a, Fbxo6a
Chromosome Location
Chr.4: 148152799 - 148160094 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_104375.1 4 - 5 647 88
GenBank mRNA BC028884.1 - 5 - 6 791 88
CX202860.1 - 4 - 5 423 88


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream F-box protein 6 Fbxo6 AA408845, FBG2, Fbs2, Fbx6b, Fbxo6b -2.2kb Sense Chr. - 148145716 - 148152135 on Build GRCm38
Downstream F-box protein 2 Fbxo2 FBG1, FBX2, Fbs1, Fbs2, NFB42, Prpl4 3.9kb Anti-sense Chr. - 148160226 - 148166417 on Build GRCm38

View Full Product Details