Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir6344
- Entrez Gene ID
-
102466629
- Gene Name
- microRNA 6344
- Gene Aliases
-
Gm27840, mmu-mir-6344
- Chromosome Location
-
Chr.1: 82131916 - 82132019 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_105762.1 |
|
1 - 1 |
45 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2 |
Nyap2 |
9430031J16Rik, Jr6, Kiaa1486 |
-766.1kb
|
Sense |
Chr. - 81076553 - 81365845 on Build GRCm38 |
Downstream |
insulin receptor substrate 1 |
Irs1 |
G972R, IRS-1 |
101.1kb
|
Anti-sense |
Chr. - 82233105 - 82291439 on Build GRCm38 |
View Full Product Details