Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rp2h
- Entrez Gene ID
-
19889
- Gene Name
- retinitis pigmentosa 2 homolog (human)
- Gene Aliases
-
AI662636, Rp2
- Chromosome Location
-
Chr.X: 20364481 - 20405653 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110967.1 |
|
4 - 5 |
1212 |
89 |
GenBank mRNA |
AK076907.1 |
- |
4 - 5 |
1074 |
89 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV270163.2 |
- |
3 - 4 |
415 |
|
Chr. X - 20377500 - 20405616 [+] on Build GRCm38 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 9 (sodium/hydrogen exchanger), member 7 |
Slc9a7 |
A530087D17Rik, NHE7 |
-105.3kb
|
Anti-sense |
Chr. - 20098851 - 20291970 on Build GRCm38 |
Downstream |
jade family PHD finger 3 |
Jade3 |
5730598B06, AI851988, AW544806, Phf16, mKIAA0215 |
22.0kb
|
Sense |
Chr. - 20425327 - 20519939 on Build GRCm38 |
View Full Product Details