Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tm6sf1
- Entrez Gene ID
-
107769
- Gene Name
- transmembrane 6 superfamily member 1
- Gene Aliases
-
AI428514, BC023123, C630016D09Rik
- Chromosome Location
-
Chr.7: 81859001 - 81884079 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111910.1 |
|
5 - 6 |
960 |
81 |
GenBank mRNA |
AK162572.1 |
- |
5 - 6 |
812 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
BTB (POZ) domain containing 1 |
Btbd1 |
1190005H08Rik |
-41.9kb
|
Anti-sense |
Chr. - 81792074 - 81829503 on Build GRCm38 |
Downstream |
hepatoma-derived growth factor, related protein 3 |
Hdgfrp3 |
2700022B06Rik, HRP-3 |
5.4kb
|
Anti-sense |
Chr. - 81881256 - 81934459 on Build GRCm38 |
View Full Product Details