Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBX1
Entrez Gene ID
101109871
Gene Name
ring-box 1
Gene Aliases
-
Chromosome Location
Chr.3: 233018425 - 233031645 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007009.3 XP_004007058.1 1 - 2 119 82

View Full Product Details