Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC9
Entrez Gene ID
443360
Gene Name
homeobox C9
Gene Aliases
HOXC-9
Chromosome Location
Chr.3: 142051006 - 142054267 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006282.4 XP_004006331.1 1 - 2 604 69
GenBank mRNA U61980.1 - 1 - 2 50 69

View Full Product Details