The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004022355.4 | XP_004022404.1 | 5 - 6 | 813 | 104 |
XM_004022356.4 | XP_004022405.1 | 5 - 6 | 670 | 104 | |
XM_012107356.3 | XP_011962746.1 | 5 - 6 | 712 | 104 | |
XM_027962993.1 | XP_027818794.1 | 5 - 6 | 853 | 104 |