Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSCL2
Entrez Gene ID
101113634
Gene Name
BSCL2 lipid droplet biogenesis associated, seipin
Gene Aliases
-
Chromosome Location
Chr.21: 42877670 - 42893152 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012102042.2 XP_011957432.1 8 - 9 1216 98
XM_012102044.3 XP_011957434.1 8 - 9 1231 98
XM_012102045.3 XP_011957435.1 8 - 9 1250 98
XM_012102047.2 XP_011957437.1 8 - 9 1585 98
XM_012102048.3 XP_011957438.1 7 - 8 984 98
XM_012102049.3 XP_011957439.1 7 - 8 1076 98
XM_027960097.1 XP_027815898.1 8 - 9 1214 98
XM_027960098.1 XP_027815899.1 8 - 9 1214 98
XM_027960099.1 XP_027815900.1 8 - 9 1214 98

View Full Product Details