Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3BGRL3
Entrez Gene ID
101111864
Gene Name
SH3 domain binding glutamate rich protein like 3
Gene Aliases
-
Chromosome Location
Chr.2: 256359973 - 256361456 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005095.4 XP_004005144.1 1 - 2 175 73

View Full Product Details