Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCLAF3
Entrez Gene ID
101110907
Gene Name
BCLAF1 and THRAP3 family member 3
Gene Aliases
CXHXorf23
Chromosome Location
Chr.X: 19013976 - 19070080 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106372.3 XP_011961762.1 7 - 8 1815 64
XM_015104819.2 XP_014960305.1 7 - 8 1800 64
XM_015104820.2 XP_014960306.1 6 - 7 1608 64
XM_015104821.2 XP_014960307.1 7 - 8 1800 64
XM_015104822.2 XP_014960308.1 7 - 8 1740 64
XM_015104824.2 XP_014960310.1 7 - 8 1800 64

View Full Product Details