Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GJD2
Entrez Gene ID
101103323
Gene Name
gap junction protein delta 2
Gene Aliases
-
Chromosome Location
Chr.7: 29957477 - 29961022 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010426.4 XP_004010475.1 1 - 2 199 82

View Full Product Details