Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA4
Entrez Gene ID
101122593
Gene Name
NDUFA4 mitochondrial complex associated
Gene Aliases
-
Chromosome Location
Chr.4: 21617013 - 21624306 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007753.3 XP_004007802.1 2 - 3 258 73

View Full Product Details