Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC6A
Entrez Gene ID
678675
Gene Name
C-type lectin domain containing 6A
Gene Aliases
-
Chromosome Location
Chr.3: 222786234 - 222800794 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006899.4 XP_004006948.1 3 - 4 446 100
XM_004006900.4 XP_004006949.1 4 - 5 543 100
GenBank mRNA AM167931.1 - 3 - 4 279 100

View Full Product Details