Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CXHXorf65
Entrez Gene ID
101103295
Gene Name
chromosome X CXorf65 homolog
Gene Aliases
-
Chromosome Location
Chr.X: 66567225 - 66569579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022162.4 XP_004022211.1 3 - 4 726 110

View Full Product Details