Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBP
Entrez Gene ID
100192428
Gene Name
D-box binding PAR bZIP transcription factor
Gene Aliases
-
Chromosome Location
Chr.14: 58729502 - 58735067 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139449.1 NP_001132921.1 3 - 4 762 67
GenBank mRNA EU293835.1 - 3 - 4 762 67

View Full Product Details