Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V1F
Entrez Gene ID
101112724
Gene Name
ATPase H+ transporting V1 subunit F
Gene Aliases
-
Chromosome Location
Chr.4: 101663754 - 101666516 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008049.4 XP_004008098.1 1 - 2 270 78

View Full Product Details