Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YEATS4
Entrez Gene ID
100302361
Gene Name
YEATS domain containing 4
Gene Aliases
-
Chromosome Location
Chr.3: 161288824 - 161311395 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163059.1 NP_001156531.1 4 - 5 332 127
GenBank mRNA FJ958351.1 - 4 - 5 332 127

View Full Product Details