Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC1
Entrez Gene ID
101112949
Gene Name
stanniocalcin 1
Gene Aliases
-
Chromosome Location
Chr.2: 44122882 - 44137567 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004182.4 XP_004004231.3 2 - 3 482 70

View Full Product Details