Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAF2
Entrez Gene ID
101106145
Gene Name
PRA1 domain family member 2
Gene Aliases
-
Chromosome Location
Chr.X: 58366767 - 58369810 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963403.1 XP_027819204.1 2 - 3 774 61

View Full Product Details