Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYCR2
Entrez Gene ID
101111821
Gene Name
pyrroline-5-carboxylate reductase 2
Gene Aliases
-
Chromosome Location
Chr.12: 31638657 - 31642872 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976310.1 XP_027832111.1 2 - 3 293 69

View Full Product Details