Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS4B
Entrez Gene ID
101102029
Gene Name
ankyrin repeat and sterile alpha motif domain containing 4B
Gene Aliases
-
Chromosome Location
Chr.24: 19583867 - 19595698 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961563.1 XP_027817364.1 1 - 2 704 78

View Full Product Details