Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT4
Entrez Gene ID
100217404
Gene Name
sirtuin 4
Gene Aliases
-
Chromosome Location
Chr.17: 71820398 - 71834794 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163044.1 NP_001156516.1 1 - 2 500 64
XM_027980224.1 XP_027836025.1 2 - 3 1090 64
XM_027980225.1 XP_027836026.1 3 - 4 604 64
XM_027980226.1 XP_027836027.1 2 - 3 557 64
GenBank mRNA FJ969402.1 - 1 - 2 500 64

View Full Product Details