Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OGFR
Entrez Gene ID
101120430
Gene Name
opioid growth factor receptor
Gene Aliases
-
Chromosome Location
Chr.13: 56926261 - 56933723 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099477.2 XP_014954963.2 4 - 5 435 79

View Full Product Details