Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXRED1
Entrez Gene ID
101114222
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.21: 30620995 - 30628899 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959361.1 XP_027815162.1 5 - 6 698 59

View Full Product Details