Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP2C
Entrez Gene ID
101111258
Gene Name
RAP2C, member of RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.X: 111294932 - 111308353 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962879.1 XP_027818680.1 3 - 4 1269 117

View Full Product Details