Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB8
Entrez Gene ID
101108118
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
-
Chromosome Location
Chr.3: 148573612 - 148582185 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006405.4 XP_004006454.1 2 - 3 269 67
XM_012174379.3 XP_012029769.1 2 - 3 271 67

View Full Product Details