Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK1A
Entrez Gene ID
101104442
Gene Name
divergent protein kinase domain 1A
Gene Aliases
FAM69A
Chromosome Location
Chr.1: 74158761 - 74283989 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002198.4 XP_004002247.1 2 - 3 247 78
XM_015092116.2 XP_014947602.2 2 - 3 276 78

View Full Product Details