Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPPP
Entrez Gene ID
101110090
Gene Name
tubulin polymerization promoting protein
Gene Aliases
-
Chromosome Location
Chr.16: 77951714 - 77972643 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027980204.1 XP_027836005.1 3 - 4 559 87

View Full Product Details